Assay Details
| Gene Symbol: LINC00385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI203837.1 | - | 1 | 161 | 80 | |
| Gene Symbol: LINC00384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648491.1 | - | 1 | 5193 | 80 | |
More Information
Set Membership: |
|

LINC00385
Genomic Map