Assay Details
| Gene Symbol: TRPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135955.1 | NP_001129427.1 | 6 | 1781 | 73 | |
| NM_001135956.1 | NP_001129428.1 | 6 | 1781 | 73 | ||
| NM_001135957.1 | NP_001129429.1 | 6 | 1781 | 73 | ||
| NM_001135958.1 | NP_001129430.1 | 5 | 1262 | 73 | ||
| NM_003306.1 | NP_003297.1 | 6 | 1781 | 73 | ||
| NM_016179.2 | NP_057263.1 | 6 | 1781 | 73 | ||
| XM_011535206.1 | XP_011533508.1 | 3 | 401 | 73 | ||
| XM_017020723.1 | XP_016876212.1 | 3 | 401 | 73 | ||
| XM_017020724.1 | XP_016876213.1 | 6 | 2342 | 73 | ||
| GenBank mRNA | AF063822.1 | - | 6 | 1781 | 73 | |
| AF063823.1 | - | 6 | 1781 | 73 | ||
| AF063824.1 | - | 6 | 1781 | 73 | ||
| AF063825.1 | - | 6 | 1781 | 73 | ||
| AF175406.1 | - | 6 | 1777 | 73 | ||
| AF421358.1 | - | 5 | 1546 | 73 | ||
| AF421359.1 | - | 5 | 1546 | 73 | ||
| AF421360.1 | - | 5 | 1546 | 73 | ||
| AF421361.1 | - | 4 | 1209 | 73 | ||
| AF421362.1 | - | 4 | 1027 | 73 | ||
| AK312524.1 | - | 6 | 1751 | 73 | ||
| BC104725.1 | - | 6 | 1702 | 40008478 | 73 | |
| BC105641.1 | - | 6 | 1702 | 40008481 | 73 | |
| BC105917.1 | - | 6 | 1702 | 40008477 | 73 | |
| BX648312.1 | - | 6 | 2405 | 73 | ||
More Information
Set Membership: |
|

TRPC4
Genomic Map