Assay Details
| Gene Symbol: DIAPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042517.1 | NP_001035982.1 | 28 | 4459 | 92 | |
| NM_001258366.1 | NP_001245295.1 | 27 | 4426 | 92 | ||
| NM_001258367.1 | NP_001245296.1 | 26 | 4321 | 92 | ||
| NM_001258368.1 | NP_001245297.1 | 26 | 4249 | 92 | ||
| XM_006719876.1 | XP_006719939.1 | 22 | 3610 | 92 | ||
| GenBank mRNA | BC041395.1 | - | 2 | 1029 | 92 | |
| CA310418.1 | - | 1 | 92 | |||
More Information
Set Membership: |
|

DIAPH3
Genomic Map