| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000554703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000554703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000554703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000554703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000298406.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000554703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000554703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000554703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000554703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000555166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
ENST00000556492.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007285.hg.1 |
NM_001011713.2 |
Refseq |