Assay Details
| Gene Symbol: SRP14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040059.1 | 4 | 2006 | 83 | ||
| NR_040060.1 | 3 | 1723 | 83 | |||
| GenBank mRNA | AK127696.1 | - | 3 | 1721 | 83 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000504245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000559012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | ENST00000560341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040061.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040062.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006931.hg.1 | NR_040062.1 | Refseq |

SRP14-AS1
Genomic Map