Assay Details
| Gene Symbol: GDAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017686.3 | NP_060156.1 | 14 | 3574 | 101 | |
| XM_005270959.2 | XP_005271016.1 | 15 | 3640 | 101 | ||
| XM_011541623.2 | XP_011539925.1 | 15 | 3670 | 101 | ||
| XM_017001531.1 | XP_016857020.1 | 16 | 3662 | 101 | ||
| XM_017001532.1 | XP_016857021.1 | 15 | 4017 | 101 | ||
| GenBank mRNA | BX647355.1 | - | 1 | 208 | 101 | |
More Information
Set Membership: |
|

GDAP2
Genomic Map