Assay Details
| Gene Symbol: DDTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011529816.2 | XP_011528118.1 | 3 | 1270 | 64 | |
| GenBank mRNA | AK296189.1 | - | 2 | 1027 | 64 | |
| AK298326.1 | - | 3 | 599 | 64 | ||
More Information
Set Membership: |
|

DDTL
Genomic Map