Assay Details
| Gene Symbol: GAREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168241.1 | NP_001161713.1 | 6 | 1809 | 65 | |
| XM_011532564.2 | XP_011530866.1 | 6 | 2246 | 65 | ||
| XM_011532565.2 | XP_011530867.1 | 7 | 1819 | 65 | ||
| XM_011532566.2 | XP_011530868.1 | 6 | 1577 | 65 | ||
| GenBank mRNA | AB124552.1 | - | 6 | 1809 | 65 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |

GAREM2
Genomic Map