Assay Details
Gene Symbol: GXYLT1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121572.1 | 1 | 1615 | 90 | ||
GenBank mRNA | DA103059.1 | - | 1 | 471 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007415.hg.1 | ENST00000456472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007415.hg.1 | ENST00000456472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013452.hg.1 | ENST00000424978.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007246.hg.1 | ENST00000453058.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007246.hg.1 | ENST00000453058.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007415.hg.1 | ENST00000456472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007415.hg.1 | ENST00000456472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013452.hg.1 | ENST00000424978.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
