Assay Details
| Gene Symbol: SLC25A25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033374.1 | 5 | 3176 | 99 | ||
| GenBank mRNA | AL833509.1 | - | 5 | 3138 | 99 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000602939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000602939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | ENST00000602939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000418747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000536815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000602939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000602939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | ENST00000602939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011622.hg.1 | NR_033374.1 | Refseq |

SLC25A25-AS1
Genomic Map