| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000484537.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000484537.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000484537.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
ENST00000484537.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000378785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000475091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000484537.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000484537.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000484537.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
ENST00000484537.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006517.hg.1 |
NM_001039211.2 |
Refseq |