Assay Details
| Gene Symbol: TTLL10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125828.1 | - | 1 | 2413 | 78 | |
| AK128833.1 | - | 2 | 1789 | 78 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000506177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000514695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | ENST00000514695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379288.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000379290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000460998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000486379.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000506177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000514695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | ENST00000514695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_001130045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006492.hg.1 | NM_153254.2 | Refseq |

TTLL10-AS1
Genomic Map