Assay Details
| Gene Symbol: LINC00709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108039.1 | 2 | 98 | |||
| GenBank mRNA | AA846132.1 | - | 1 | 274 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00709| Gene Symbol: LINC00709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108039.1 | 2 | 98 | |||
| GenBank mRNA | AA846132.1 | - | 1 | 274 | 98 | |
Set Membership: |
|