Assay Details
| Gene Symbol: BAIAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199096.1 | NP_001186025.1 | 14 | 1405 | 52 | |
| NM_001199097.1 | NP_001186026.1 | 15 | 1455 | 52 | ||
| NM_001199098.1 | NP_001186027.1 | 15 | 1386 | 52 | ||
| NM_001199099.1 | NP_001186028.1 | 15 | 1371 | 52 | ||
| NM_001286464.1 | NP_001273393.1 | 15 | 1506 | 52 | ||
| NM_003933.4 | NP_003924.2 | 15 | 1563 | 52 | ||
| XM_011522728.1 | XP_011521030.1 | 15 | 2109 | 52 | ||
| XM_011522729.1 | XP_011521031.1 | 15 | 2109 | 52 | ||
| XM_011522730.2 | XP_011521032.1 | 15 | 2109 | 52 | ||
| GenBank mRNA | AB018277.2 | - | 15 | 1415 | 52 | |
| AK294576.1 | - | 4 | 326 | 52 | ||
| AK294744.1 | - | 15 | 1331 | 52 | ||
| AK295645.1 | - | 15 | 1345 | 52 | ||
| AK299309.1 | - | 15 | 1415 | 52 | ||
| AK299358.1 | - | 14 | 1405 | 52 | ||
| AK315059.1 | - | 15 | 1563 | 52 | ||
| AL834321.1 | - | 15 | 1457 | 52 | ||
| BC104966.1 | - | 15 | 1426 | 8143969 | 52 | |
| BC112129.1 | - | 15 | 1438 | 8327597 | 52 | |
More Information
Set Membership: |
|

BAIAP3
Genomic Map