Assay Details
| Gene Symbol: SMYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052928.2 | NP_443160.2 | 5 | 1111 | 88 | |
| XM_011523632.2 | XP_011521934.1 | 5 | 1092 | 88 | ||
| XM_011523633.2 | XP_011521935.1 | 5 | 1092 | 88 | ||
| XM_017024121.1 | XP_016879610.1 | 6 | 1356 | 88 | ||
| GenBank mRNA | AB067523.1 | - | 1 | 355 | 88 | |
| AK095369.1 | - | 5 | 1111 | 88 | ||
| BC035077.2 | - | 5 | 1030 | 5260399 | 88 | |
| CR987176.1 | - | 2 | 418 | 88 | ||
More Information
Set Membership: |
|

SMYD4
Genomic Map