Assay Details
Gene Symbol: FAM231B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_016999988.1 | XP_016855477.1 | 2 | 1939 | 124 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_016999988.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
