Assay Details
| Gene Symbol: SMIM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164824.1 | NP_001158296.1 | 3 | 4911 | 81 | |
| NM_001164825.1 | NP_001158297.1 | 2 | 4804 | 81 | ||
| NM_001320261.1 | NP_001307190.1 | 2 | 4686 | 81 | ||
| NM_138428.5 | NP_612437.3 | 2 | 4661 | 81 | ||
More Information
Set Membership: |
|

SMIM12
Genomic Map