Assay Details
| Gene Symbol: CCDC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005436.4 | NP_005427.2 | 2 | 626 | 94 | |
| GenBank mRNA | AK292593.1 | - | 2 | 626 | 94 | |
| BC022284.1 | - | 2 | 520 | 94 | ||
| BC036757.2 | - | 2 | 465 | 5549623 | 94 | |
| BC064391.1 | - | 2 | 452 | 5745698 | 94 | |
| S72869.1 | - | 2 | 430 | 94 | ||
More Information
Set Membership: |
|

CCDC6
Genomic Map