Assay Details
| Gene Symbol: TXNIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006472.5 | NP_006463.3 | 1 | 192 | 158 | |
| XM_017000085.1 | XP_016855574.1 | 1 | 141 | 158 | ||
| GenBank mRNA | AK226090.1 | - | 1 | 104 | 158 | |
| AK301203.1 | - | 1 | 140 | 158 | ||
| BX537824.1 | - | 1 | 140 | 158 | ||
| DB058019.1 | - | 1 | 168 | 158 | ||
| HY177853.1 | - | 1 | 194 | 159 | ||
| S73591.1 | - | 1 | 48 | 158 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK301203.1, NM_006472.5, S73591.1, XM_017000085.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
protein import into nucleus
cell cycle
response to mechanical stimulus
response to glucose
keratinocyte differentiation
response to estradiol
response to progesterone
regulation of cell proliferation
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
negative regulation of catalytic activity
platelet-derived growth factor receptor signaling pathway
response to calcium ion
negative regulation of cell division
cellular response to tumor cell
enzyme inhibitor activity
protein binding
ubiquitin protein ligase binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
protein import into nucleus
cell cycle
response to mechanical stimulus
response to glucose
keratinocyte differentiation
response to estradiol
response to progesterone
regulation of cell proliferation
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
negative regulation of catalytic activity
platelet-derived growth factor receptor signaling pathway
response to calcium ion
negative regulation of cell division
cellular response to tumor cell
enzyme inhibitor activity
protein binding
ubiquitin protein ligase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000425134.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000486597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000488537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | ENST00000582401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015598.hg.1 | NM_006472.5 | Refseq |

TXNIP
Genomic Map