Assay Details
| Gene Symbol: PROK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032414.2 | NP_115790.1 | 3 | 1210 | 86 | |
| GenBank mRNA | AF333024.1 | - | 3 | 1212 | 86 | |
| AY029225.1 | - | 3 | 1207 | 86 | ||
| AY358683.1 | - | 3 | 1249 | 86 | ||
| BC025399.1 | - | 3 | 1187 | 4826496 | 86 | |
More Information
Set Membership: |
|

PROK1
Genomic Map