Assay Details
| Gene Symbol: XCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509865.2 | XP_011508167.1 | 1 | 1056 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011509865.2 transcripts.
Set Membership: |
|

XCL1
Genomic Map