| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000367099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000367099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000367099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000367099.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000471071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000471071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000471071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
ENST00000471071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000367099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000367099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000367099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000367099.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000423557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000471071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000471071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000471071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
ENST00000471071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017107.hg.1 |
NM_000572.2 |
Refseq |