Assay Details
| Gene Symbol: LOC100287049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BF439213.1 | - | 1 | 41 | 90 | |
| Gene Symbol: DCAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK026213.1 | - | 1 | 90 | ||
More Information
Set Membership: |
|

LOC100287049
Genomic Map