Assay Details
Gene Symbol: BRINP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011509826.1 | XP_011508128.1 | 1 | 577 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011509826.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000460161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000460161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000460161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000361539.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000460161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000460161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000460161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | ENST00000478325.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010720.hg.1 | NM_021165.3 | Refseq |
