Assay Details
| Gene Symbol: LOC642852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026943.1 | 3 | 2830 | 75 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000400362.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000433465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | ENST00000454115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007412.hg.1 | NR_026943.1 | Refseq |

LOC642852
Genomic Map