Assay Details
| Gene Symbol: LOC101928107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109961.1 | 3 | 1227 | 76 | ||
| GenBank mRNA | BC021745.1 | - | 4 | 1232 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928107| Gene Symbol: LOC101928107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109961.1 | 3 | 1227 | 76 | ||
| GenBank mRNA | BC021745.1 | - | 4 | 1232 | 76 | |
Set Membership: |
|