Assay Details
Gene Symbol: CCDC115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124405.1 | - | 1 | 1315 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000259229.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000409127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000442217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | ENST00000465315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NM_032357.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014260.hg.1 | NR_104471.1 | Refseq |
