Assay Details
| Gene Symbol: FER1L5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011512124.2 | XP_011510426.1 | 1 | 549 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011512124.2 transcripts.
Set Membership: |
|

FER1L5
Genomic Map