Assay Details
| Gene Symbol: NAP1L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005969.3 | NP_005960.1 | 16 | 1552 | 118 | |
| GenBank mRNA | AK093928.1 | - | 3 | 1297 | 118 | |
| AK095320.1 | - | 2 | 3033 | 118 | ||
| AK311098.1 | - | 1 | 1064 | 118 | ||
| AK316242.1 | - | 15 | 1431 | 118 | ||
| AK316324.1 | - | 15 | 1452 | 118 | ||
| AK316548.1 | - | 16 | 1571 | 118 | ||
| BC022090.2 | - | 16 | 1462 | 3029455 | 118 | |
| U77456.1 | - | 16 | 1560 | 118 | ||
More Information
Set Membership: |
|

NAP1L4
Genomic Map