Assay Details
| Gene Symbol: PQLC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024016.2 | 3 | 724 | 78 | ||
| GenBank mRNA | AI377021.1 | - | 1 | 78 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |

PQLC2L
Genomic Map