Assay Details
| Gene Symbol: MTMR14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077525.2 | NP_001070993.1 | 17 | 1686 | 96 | |
| NM_001077526.2 | NP_001070994.1 | 17 | 1686 | 96 | ||
| XM_005265400.1 | XP_005265457.1 | 16 | 1584 | 96 | ||
| XM_011534012.1 | XP_011532314.1 | 16 | 1455 | 96 | ||
| XM_011534013.1 | XP_011532315.1 | 16 | 1411 | 96 | ||
| XM_017007034.1 | XP_016862523.1 | 18 | 1856 | 96 | ||
| XM_017007035.1 | XP_016862524.1 | 17 | 1781 | 96 | ||
| XM_017007036.1 | XP_016862525.1 | 18 | 1855 | 96 | ||
| XM_017007037.1 | XP_016862526.1 | 17 | 1781 | 96 | ||
| XM_017007038.1 | XP_016862527.1 | 16 | 1584 | 96 | ||
| XM_017007041.1 | XP_016862530.1 | 15 | 1380 | 96 | ||
| XM_017007043.1 | XP_016862532.1 | 16 | 1455 | 96 | ||
| XM_017007044.1 | XP_016862533.1 | 15 | 1502 | 96 | ||
| XM_017007045.1 | XP_016862534.1 | 15 | 1491 | 96 | ||
| GenBank mRNA | AF289603.1 | - | 17 | 1650 | 96 | |
| AK074792.1 | - | 17 | 1657 | 96 | ||
| AY545552.1 | - | 4 | 329 | 96 | ||
| BC025952.1 | - | 17 | 1636 | 4876556 | 96 | |
| BC035690.1 | - | 17 | 1598 | 5755887 | 96 | |
| BC050626.1 | - | 17 | 1609 | 4416416 | 96 | |
| DQ630520.1 | - | 17 | 1535 | 96 | ||
More Information
Set Membership: |
|

MTMR14
Genomic Map