Assay Details
| Gene Symbol: C11orf95-RELA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_C11orf95-RELA_C3R2.0 | - | 57 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11orf95-RELA fusion| Gene Symbol: C11orf95-RELA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_C11orf95-RELA_C3R2.0 | - | 57 | 108 | ||
Set Membership: |
|