Assay Details
| Gene Symbol: DCTN1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_DCTN1-MET_D26M15.0 | - | 123 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCTN1-MET fusion| Gene Symbol: DCTN1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_DCTN1-MET_D26M15.0 | - | 123 | 76 | ||