Assay Details
| Gene Symbol: EPS15-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EPS15-NTRK1_E21N9.0 | - | 118 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPS15-NTRK1 fusion| Gene Symbol: EPS15-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EPS15-NTRK1_E21N9.0 | - | 118 | 100 | ||