Assay Details
| Gene Symbol: ERC1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ERC1-PDGFRB_E15P10.0 | - | 115 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERC1-PDGFRB fusion| Gene Symbol: ERC1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ERC1-PDGFRB_E15P10.0 | - | 115 | 101 | ||
Set Membership: |
|