Assay Details
| Gene Symbol: FIP1L1-PDGFRA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FIP1L1-PDGFRA_F9P12del38.0 | - | 55 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FIP1L1-PDGFRA fusion| Gene Symbol: FIP1L1-PDGFRA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FIP1L1-PDGFRA_F9P12del38.0 | - | 55 | 91 | ||