Assay Details
| Gene Symbol: LRRFIP1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_LRRFIP1-MET_L19M15.0 | - | 121 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRFIP1-MET fusion| Gene Symbol: LRRFIP1-MET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_LRRFIP1-MET_L19M15.0 | - | 121 | 103 | ||
Set Membership: |
|