Assay Details
| Gene Symbol: LYN-NTRK3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_LYN-NTRK3_L8N14.0 | - | 122 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYN-NTRK3 fusion| Gene Symbol: LYN-NTRK3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_LYN-NTRK3_L8N14.0 | - | 122 | 101 | ||
Set Membership: |
|