Assay Details
| Gene Symbol: MYB-NFIB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYB-NFIB_M13N11_COSF876.0 | - | 118 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB-NFIB fusion| Gene Symbol: MYB-NFIB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYB-NFIB_M13N11_COSF876.0 | - | 118 | 108 | ||
Set Membership: |
|