Assay Details
| Gene Symbol: PDE7A-NRG1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PDE7A-NRG1_P3N6.0 | - | 121 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDE7A-NRG1 fusion| Gene Symbol: PDE7A-NRG1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PDE7A-NRG1_P3N6.0 | - | 121 | 94 | ||