Assay Details
| Gene Symbol: AP3B1-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AP3B1-BRAF_A22B9.0 | - | 63 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AP3B1-BRAF fusion| Gene Symbol: AP3B1-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AP3B1-BRAF_A22B9.0 | - | 63 | 95 | ||