Assay Details
| Gene Symbol: AR-AR fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AR-AR_A6A8_V13.0 | - | 65 | 88 | ||
| OM_AR-AR_A6A9_V18.0 | - | 65 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AR-AR fusion| Gene Symbol: AR-AR fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_AR-AR_A6A8_V13.0 | - | 65 | 88 | ||
| OM_AR-AR_A6A9_V18.0 | - | 65 | 95 | |||