Assay Details
| Gene Symbol: CLCN6-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CLCN6-RAF1_C2R8.0 | - | 62 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLCN6-RAF1 fusion| Gene Symbol: CLCN6-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CLCN6-RAF1_C2R8.0 | - | 62 | 86 | ||