Assay Details
| Gene Symbol: CTNNB1-FGFR2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CTNNB1-FGFR2_C1F10.0 | - | 66 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTNNB1-FGFR2 fusion| Gene Symbol: CTNNB1-FGFR2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CTNNB1-FGFR2_C1F10.0 | - | 66 | 86 | ||