Assay Details
| Gene Symbol: EBF1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EBF1-PDGFRB_E11P11.0 | - | 50 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EBF1-PDGFRB fusion| Gene Symbol: EBF1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EBF1-PDGFRB_E11P11.0 | - | 50 | 89 | ||