Assay Details
| Gene Symbol: HIP1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_HIP1-PDGFRB_H30P11.0 | - | 50 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIP1-PDGFRB fusion| Gene Symbol: HIP1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_HIP1-PDGFRB_H30P11.0 | - | 50 | 95 | ||