Assay Details
| Gene Symbol: KANK1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KANK1-PDGFRB_K2P9.0 | - | 67 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KANK1-PDGFRB fusion| Gene Symbol: KANK1-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KANK1-PDGFRB_K2P9.0 | - | 67 | 88 | ||