Assay Details
| Gene Symbol: MDM4-MDM4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MDM4-MDM4_M3M10.0 | - | 54 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MDM4-MDM4 fusion| Gene Symbol: MDM4-MDM4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MDM4-MDM4_M3M10.0 | - | 54 | 115 | ||
Set Membership: |
|