Assay Details
| Gene Symbol: SLC12A7-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SLC12A7-BRAF_S17B11.0 | - | 67 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC12A7-BRAF fusion| Gene Symbol: SLC12A7-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SLC12A7-BRAF_S17B11.0 | - | 67 | 71 | ||