Assay Details
| Gene Symbol: TPM3-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TPM3-NTRK1_T7N13.0 | - | 45 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPM3-NTRK1 fusion| Gene Symbol: TPM3-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TPM3-NTRK1_T7N13.0 | - | 45 | 85 | ||